Boc-N-methyl-D-alanine ≥95%

Supplier: Matrix Scientific

Synonyms: Boc-N-Me-Ala-OH, N-Boc-N-methyl-L-alanine

042436-5G 042436-25G
101855-794EA 56.92 USD
101855-794 101855-796
Boc-N-methyl-D-alanine ≥95%
Boc-N-methyl-D-alanine

Hazard: Irritant

Formula: C₉H₁₇NO₄
MW: 203.2 g/mol
Melting Pt: 88…92 °C
MDL Number: MFCD00037242
CAS Number: 16948-16-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR