Boc-N-methyl-D-Valine

Supplier: Matrix Scientific

Synonyms: Boc-D-N-Me-Val-OH, N-Boc-N-methyl-D-valine, (R)-2-((tert-Butoxycarbonyl)(methyl)amino)-3-methylbutanoic acid, Boc-N-Me-D-Val-OH

042434-1G 042434-5G
101855-786EA 53.85 USD
101855-786 101855-788
Boc-N-methyl-D-Valine
Boc-N-methyl-D-Valine

Hazard: Irritant

Formula: C₁₁H₂₁NO₄
MW: 231.292 g/mol
MDL Number: MFCD00076996
CAS Number: 89536-85-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR