Boc-Nα-methyl-D-tryptophan

Supplier: Matrix Scientific

042433-1G 042433-5G
101855-782EA 661.54 USD
101855-782 101855-784
Boc-Nα-methyl-D-tryptophan
Boc-Nalpha-methyl-D-tryptophan

Hazard: Irritant

Formula: C₁₇H₂₂N₂O₄
MDL Number: MFCD04974457
CAS Number: 142381-30-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR