Boc-N-α-methyl-D-phenylalanine . DCHA
Supplier: Matrix Scientific
Synonyms:
Boc-D-N-Me-Phe DCHA, Boc-D-N-Me-Phe-OH·DCHA
042432-1G
042432-5G
101855-778EA
498.46
USD
101855-778
101855-780
Boc-N-α-methyl-D-phenylalanine . DCHA
Boc-N-alpha-methyl-D-phenylalanine . DCHA
Hazard: Irritant
Formula:
C₁₅H₂₁NO₄·C₁₂H₂₃N MW: 460.66 g/mol |
MDL Number:
MFCD00058079 CAS Number: 102185-45-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...