Boc-Nα-methyl-DL-tryptophan

Supplier: Matrix Scientific

042431-100MG 042431-1G
101855-774EA 263.08 USD
101855-774 101855-776
Boc-Nα-methyl-DL-tryptophan
Boc-Nalpha-methyl-DL-tryptophan

Hazard: Irritant

Formula: C₁₇H₂₂N₂O₄
MW: 318.38 g/mol
MDL Number: MFCD02682370
CAS Number: 1452574-00-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR