Boc-Nα-methyl-D-glutamic acid γ-benzylester

Supplier: Matrix Scientific

042429-1G 042429-5G
101855-766EA 823.08 USD
101855-766 101855-768
Boc-Nα-methyl-D-glutamic acid γ-benzylester
Boc-Nalpha-methyl-D-glutamic acid gamma-benzylester

Hazard: Irritant

Formula: C₁₈H₂₅NO₆
MDL Number: MFCD04974253
CAS Number: 1562452-07-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR