Boc-Nα-methyl-4-chloro-L-phenylalanine
Supplier: Matrix Scientific
Synonyms:
(S)-2-((tert-Butoxycarbonyl)(methyl)amino)-3-(4-chlorophenyl)propanoic acid, Boc-N-Me-Phe(4-Cl)-OH
042427-1G
042427-5G
101855-758EA
598.46
USD
101855-758
101855-760
Boc-Nα-methyl-4-chloro-L-phenylalanine
Boc-Nalpha-methyl-4-chloro-L-phenylalanine
Hazard: Irritant
Formula:
C₁₅H₂₀ClNO₄ MW: 313.78 g/mol |
MDL Number:
MFCD00672517 CAS Number: 179033-68-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...