Boc-Nα-methyl-4-chloro-D-phenylalanine

Supplier: Matrix Scientific

Synonyms: Boc-N-Me-p-chloro-D-Phe-OH

042426-250MG 042426-1G
101855-754EA 300 USD
101855-754 101855-756
Boc-Nα-methyl-4-chloro-D-phenylalanine
Boc-Nalpha-methyl-4-chloro-D-phenylalanine

Hazard: Irritant

Formula: C₁₅H₂₀ClNO₄
MW: 313.778 g/mol
MDL Number: MFCD00236864
CAS Number: 125324-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR