Boc-N-Me-Arg(Mtr)-OH

Supplier: Matrix Scientific

042425-1G 042425-5G
101855-750EA 547.69 USD
101855-750 101855-752
Boc-N-Me-Arg(Mtr)-OH
Boc-N-Me-Arg(Mtr)-OH

Hazard: Irritant

Formula: C₂₂H₃₆N₄O₇S
MW: 500.611 g/mol
MDL Number: MFCD00236863
CAS Number: 125602-26-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR