N-(1-Boc-piperidine-4-yl)-L-proline

Supplier: Matrix Scientific

042423-1G 042423-5G
101855-746EA 300 USD
101855-746 101855-748
N-(1-Boc-piperidine-4-yl)-L-proline
N-(1-Boc-piperidine-4-yl)-L-proline

Hazard: Irritant

Formula: C₁₅H₂₆N₂O₄
MDL Number: MFCD01861211
CAS Number: 221352-39-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR