Fmoc-trans-3-azabicyclo[3.1.0]hexane-2-carboxylicacid

Supplier: Matrix Scientific

042422-250MG 042422-1G
101855-742EA 823.08 USD
101855-742 101855-744
Fmoc-trans-3-azabicyclo[3.1.0]hexane-2-carboxylicacid
Fmoc-trans-3-azabicyclo[3.1.0]hexane-2-carboxylicacid

Hazard: Irritant

Formula: C₂₁H₁₉NO₄
MDL Number: MFCD02682401
CAS Number: 1219181-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR