Fmoc-4-phenylpiperidine-4-carboxylic acid

Supplier: Matrix Scientific

042418-1G 042418-5G
101855-726EA 138.46 USD
101855-726 101855-728
Fmoc-4-phenylpiperidine-4-carboxylic acid
Fmoc-4-phenylpiperidine-4-carboxylic acid

Hazard: Irritant

Formula: C₂₇H₂₅NO₄
MDL Number: MFCD00673787
CAS Number: 215190-19-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR