Fmoc-4-amino-(1-carboxymethyl) piperidine
Supplier: Matrix Scientific
042417-1G
042417-5G
101855-722EA
400
USD
101855-722
101855-724
Fmoc-4-amino-(1-carboxymethyl) piperidine
Fmoc-4-amino-(1-carboxymethyl) piperidine
Hazard: Irritant
Formula:
C₂₂H₂₄N₂O₄ |
MDL Number:
MFCD01321021 CAS Number: 221352-82-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...