Fmoc-4-(2-aminoethyl)(1-carboxymethyl)piperazine dihydrochloride

Supplier: Matrix Scientific

Synonyms: 2-(4-(2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)ethyl)piperazin-1-yl)acetic acid dihydrochloride, 4-[2-(Fmoc-amino)ethyl]piperazin-1-ylacetic acid dihydrochloride

042416-1G 042416-5G
101855-718EA 326.15 USD
101855-718 101855-720
Fmoc-4-(2-aminoethyl)(1-carboxymethyl)piperazine dihydrochloride
Fmoc-4-(2-aminoethyl)(1-carboxymethyl)piperazine dihydrochloride

Hazard: Irritant

Formula: C₂₃H₂₉Cl₂N₃O₄
MDL Number: MFCD00673781
CAS Number: 204320-65-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR