Fmoc-3-carboxypiperidine

Supplier: Matrix Scientific

Synonyms: 1-(((9H-Fluoren-9-yl)methoxy)carbonyl)piperidine-3-carboxylic acid

042415-1G 042415-5G
101855-714EA 224.62 USD
101855-714 101855-716
Fmoc-3-carboxypiperidine
Fmoc-3-carboxypiperidine

Hazard: Irritant

Formula: C₂₁H₂₁NO₄
MDL Number: MFCD00673789
CAS Number: 158922-07-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR