Fmoc-2-aminothiazole-4-acetic acid

Supplier: Matrix Scientific

042413-1G 042413-5G
101855-706EA 300 USD
101855-706 101855-708
Fmoc-2-aminothiazole-4-acetic acid
Fmoc-2-aminothiazole-4-acetic acid

Hazard: Irritant

Formula: C₂₀H₁₆N₂O₄S
MDL Number: MFCD02682395
CAS Number: 1185300-16-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR