Fmoc-1-aminoindan-1-carboxylic acid

Supplier: Matrix Scientific

Synonyms: N-FMOC-DL-1-Aminoindane-1-carboxylic acid

042409-1G 042409-5G
101855-690EA 412.31 USD
101855-690 101855-692
Fmoc-1-aminoindan-1-carboxylic acid
Fmoc-1-aminoindan-1-carboxylic acid

Hazard: Irritant

Formula: C₂₅H₂₁NO₄
MW: 399.45 g/mol
MDL Number: MFCD01321019
CAS Number: 214139-28-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR