(S)-1-[(9H-Fluoren-9-ylmethoxy)carbonyl]-2-piperidinecarboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)piperidine-2-carboxylic acid, Fmoc-Pip-OH, 1-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-pipecolic Acid, Fmoc-HoPro-OH, 1-Fmoc-L-pipecolic Acid, Fmoc-HomoPro-OH, (S)-1-Fmoc-2-piperidinecarboxylic Acid

042406-1G 042406-5G
101855-678EA 29.23 USD
101855-678 101855-680
(S)-1-[(9H-Fluoren-9-ylmethoxy)carbonyl]-2-piperidinecarboxylic acid ≥95%
(S)-1-[(9H-Fluoren-9-ylmethoxy)carbonyl]-2-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₂₁H₂₁NO₄
MW: 351.40 g/mol
Melting Pt: 153…158 °C
MDL Number: MFCD00235898
CAS Number: 86069-86-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR