N-Fmoc-DL-pipecolic acid

Supplier: Matrix Scientific

Synonyms: 1-(((9H-Fluoren-9-yl)methoxy)carbonyl)piperidine-2-carboxylic acid

042405-1G 042405-5G
101855-674EA 224.62 USD
101855-674 101855-676
N-Fmoc-DL-pipecolic acid
N-Fmoc-DL-pipecolic acid

Hazard: Irritant

Formula: C₂₁H₂₁NO₄
MW: 351.402 g/mol
MDL Number: MFCD01823265
CAS Number: 105751-19-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR