Fmoc-(R)-nipecotic acid

Supplier: Matrix Scientific

Synonyms: (R)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)piperidine-3-carboxylic acid

042404-1G 042404-5G
101855-670EA 275.38 USD
101855-670 101855-672
Fmoc-(R)-nipecotic acid
Fmoc-(R)-nipecotic acid

Hazard: Irritant

Formula: C₂₁H₂₁NO₄
MDL Number: MFCD04112683
CAS Number: 193693-67-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR