Fmoc-d-pipecolic acid

Supplier: Matrix Scientific

Synonyms: (R)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)piperidine-2-carboxylic acid, 1-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-pipecolic acid, (R)-1-Fmoc-2-piperidinecarboxylic acid, Fmoc-D-Pip-OH, 1-Fmoc-D-pipecolic acid, Fmoc-D-Homopro-OH, (R)-1-[(9H-Fluoren-9-ylmethoxy)carbonyl]-2-piperidinecarboxylic acid

042403-1G 042403-5G
101855-666EA 106.15 USD
101855-666 101855-668
Fmoc-d-pipecolic acid
Fmoc-d-pipecolic acid

Hazard: Irritant

Formula: C₂₁H₂₁NO₄
MW: 351.402 g/mol
MDL Number: MFCD00235899
CAS Number: 101555-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR