Boc-3-azabicyclo[3.1.0]hexane-2-carboxylicacid
Supplier: Matrix Scientific
042401-250MG
042401-1G
101855-658EA
910.77
USD
101855-658
101855-660
Boc-3-azabicyclo[3.1.0]hexane-2-carboxylicacid
Boc-3-azabicyclo[3.1.0]hexane-2-carboxylicacid
Hazard: Irritant
Formula:
C₁₁H₁₇NO₄ |
MDL Number:
MFCD09038259 CAS Number: 953061-58-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...