1-Boc-piperidine-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(tert-Butoxycarbonyl)piperidine-4-carboxylic acid, N-BOC-4-Piperidinecarboxylic acid, Boc-Inp-OH, 1-Boc-isonipecotic acid

042400-25G 042400-100G
101855-654EA 43.08 USD
101855-654 101855-656
1-Boc-piperidine-4-carboxylic acid ≥95%
1-Boc-piperidine-4-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₉NO₄
MW: 229.3 g/mol
Melting Pt: 149-151°C
MDL Number: MFCD00076999
CAS Number: 84358-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR