Boc-L-indoline-2-carboxylic acid

Supplier: Matrix Scientific

Synonyms: (S)-1-(tert-Butoxycarbonyl)indoline-2-carboxylic acid

042399-1G 042399-5G
101855-648EA 326.15 USD
101855-648 101855-650
Boc-L-indoline-2-carboxylic acid
Boc-L-indoline-2-carboxylic acid

Hazard: Irritant

Formula: C₁₄H₁₇NO₄
MW: 263.3 g/mol
MDL Number: MFCD00273433
CAS Number: 144069-67-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR