Boc-4-amino-(1-carboxymethyl)piperidine

Supplier: Matrix Scientific

042397-1G 042397-5G
101855-640EA 484.62 USD
101855-640 101855-642
Boc-4-amino-(1-carboxymethyl)piperidine
Boc-4-amino-(1-carboxymethyl)piperidine

Hazard: Irritant

Formula: C₁₂H₂₂N₂O₄
MW: 258.32 g/mol
MDL Number: MFCD09750500
CAS Number: 299203-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR