1-Boc-3-piperidineacetic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Boc-Piperidine-3-acetic acid

042393-1G 042393-5G
101855-628EA 38.46 USD
101855-628 101855-630
1-Boc-3-piperidineacetic acid ≥95%
1-Boc-3-piperidineacetic acid

Hazard: Irritant

Formula: C₁₂H₂₁NO₄
MW: 243.3000 g/mol
MDL Number: MFCD02178999
CAS Number: 183483-09-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR