Boc-1-aminoindan-1-carboxylic acid

Supplier: Matrix Scientific

Synonyms: 1-(Boc-amino)-1-indanecarboxylic Acid

042392-1G 042392-5G
101855-624EA 412.31 USD
101855-624 101855-626
Boc-1-aminoindan-1-carboxylic acid
Boc-1-aminoindan-1-carboxylic acid

Hazard: Irritant

Formula: C₁₅H₁₉NO₄
MW: 277.32 g/mol
MDL Number: MFCD00800596
CAS Number: 214139-26-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR