Boc-1-amino-1,2,3,4-tetrahydronaphthalene-1-carboxylic acid

Supplier: Matrix Scientific

042391-1G 042391-5G
101855-620EA 823.08 USD
101855-620 101855-622
Boc-1-amino-1,2,3,4-tetrahydronaphthalene-1-carboxylic acid
Boc-1-amino-1,2,3,4-tetrahydronaphthalene-1-carboxylic acid

Hazard: Irritant

Formula: C₁₆H₂₁NO₄
MW: 291.35 g/mol
MDL Number: MFCD04115523
CAS Number: 898404-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR