(S)-1-Boc-piperidine-3-carboxylic acid

Supplier: Matrix Scientific

Synonyms: (S)-Boc-nipecotic acid, 1-Boc-L-nipecotic acid, (S)-1-(tert-Butoxycarbonyl)piperidine-3-carboxylic acid

042390-1G 042390-5G
101855-616EA 15.38 USD
101855-616 101855-618
(S)-1-Boc-piperidine-3-carboxylic acid
(S)-1-Boc-piperidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₉NO₄
MW: 229.28 g/mol
MDL Number: MFCD02179172
CAS Number: 88495-54-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR