(R)-1-Boc-piperidine-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: (R)-1-(tert-Butoxycarbonyl)piperidine-3-carboxylic acid, Boc-D-Nip-OH, 1-Boc-D-nipecotic acid, (R)-N-BOC-piperidine-3-carboxylic acid

042387-1G 042387-5G
101855-604EA 169.23 USD
101855-604 101855-606
(R)-1-Boc-piperidine-3-carboxylic acid ≥95%
(R)-1-Boc-piperidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₉NO₄
MW: 229.28 g/mol
MDL Number: MFCD02179173
CAS Number: 163438-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR