(R)-1-Boc-piperidine-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: (R)-(+)-N-BOC-2-Piperidinecarboxylic acid, Boc-D-Homopro-OH, Boc-D-HoPro-OH, (R)-1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid

042386-5G 042386-25G
101855-600EA 36.92 USD
101855-600 101855-602
(R)-1-Boc-piperidine-2-carboxylic acid ≥95%
(R)-1-Boc-piperidine-2-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₉NO₄
MDL Number: MFCD00237380
CAS Number: 28697-17-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR