Boc-(4-carboxymethyl)piperazine ≥95%

Supplier: Matrix Scientific

Synonyms: [1-(tert-Butoxycarbonyl)piperazin-4-yl]acetic acid, 2-(4-(tert-Butoxycarbonyl)piperazin-1-yl)acetic acid

042385-1G 042385-5G
101855-596EA 60 USD
101855-596 101855-598
Boc-(4-carboxymethyl)piperazine ≥95%
Boc-(4-carboxymethyl)piperazine

Hazard: Irritant

Formula: C₁₁H₂₀N₂O₄
MDL Number: MFCD02682402
CAS Number: 156478-71-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR