4-(1-Fmoc-piperidine-4-yl)butanoic acid

Supplier: Matrix Scientific

042384-1G 042384-5G
101855-592EA 143.08 USD
101855-592 101855-594
4-(1-Fmoc-piperidine-4-yl)butanoic acid
4-(1-Fmoc-piperidine-4-yl)butanoic acid

Hazard: Irritant

Formula: C₂₄H₂₇NO₄
MDL Number: MFCD04114964
CAS Number: 885274-47-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR