4-(1-Boc-piperidine-4-yl)butanoic acid

Supplier: Matrix Scientific

Synonyms: 4-(1-(tert-Butoxycarbonyl)piperidin-4-yl)butanoic acid

042383-1G 042383-5G
101855-588EA 338.46 USD
101855-588 101855-590
4-(1-Boc-piperidine-4-yl)butanoic acid
4-(1-Boc-piperidine-4-yl)butanoic acid

Hazard: Irritant

Formula: C₁₄H₂₅NO₄
MW: 271.36 g/mol
MDL Number: MFCD04114965
CAS Number: 142247-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR