3-(1-Fmoc-piperidine-4-yl)propionic acid
Supplier: Matrix Scientific
Synonyms:
3-(1-(((9H-Fluoren-9-yl)methoxy)carbonyl)piperidin-4-yl)propanoic acid
042382-1G
042382-5G
101855-584EA
573.85
USD
101855-584
101855-586
3-(1-Fmoc-piperidine-4-yl)propionic acid
3-(1-Fmoc-piperidine-4-yl)propionic acid
Hazard: Irritant
Formula:
C₂₃H₂₅NO₄ MW: 379.46 g/mol |
MDL Number:
MFCD02179125 CAS Number: 154938-68-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...