3-(1-Boc-piperidine-4-yl)propionic acid

Supplier: Matrix Scientific

042381-1G 042381-5G
101855-580EA 150.77 USD
101855-580 101855-582
3-(1-Boc-piperidine-4-yl)propionic acid
3-(1-Boc-piperidine-4-yl)propionic acid

Hazard: Irritant

Formula: C₁₃H₂₃NO₄
MDL Number: MFCD01861212
CAS Number: 154375-43-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR