Fmoc-O-t-butyl-L-β-homotyrosine

Supplier: Matrix Scientific

Synonyms: Fmoc-β-HoTyr(tBu)-OH, Fmoc-β-HomoTyr(tBu)-OH

042380-250MG 042380-1G
101855-576EA 349.23 USD
101855-576 101855-578
Fmoc-O-t-butyl-L-β-homotyrosine
Fmoc-O-t-butyl-L-beta-homotyrosine

Hazard: Irritant

Formula: C₂₉H₃₁NO₅
MW: 473.57 g/mol
MDL Number: MFCD01862862
CAS Number: 219967-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR