Fmoc-O-t-butyl-L-β-homoserine

Supplier: Matrix Scientific

Synonyms: Fmoc-β-HomoSer(tBu)-OH, Fmoc-β-HoSer(tBu)-OH

042378-250MG 042378-1G
101855-568EA 349.23 USD
101855-568 101855-570
Fmoc-O-t-butyl-L-β-homoserine
Fmoc-O-t-butyl-L-beta-homoserine

Hazard: Irritant

Formula: C₂₃H₂₇NO₅
MW: 397.48 g/mol
MDL Number: MFCD01862865
CAS Number: 203854-51-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR