Fmoc-O-t-butyl-L-β-homohydroxyproline

Supplier: Matrix Scientific

Synonyms: 2-((2S,4R)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)-4-(tert-butoxy)pyrrolidin-2-yl)acetic acid

042377-100MG 042377-500MG
101855-564EA 364.62 USD
101855-564 101855-566
Fmoc-O-t-butyl-L-β-homohydroxyproline
Fmoc-O-t-butyl-L-beta-homohydroxyproline

Hazard: Irritant

Formula: C₂₅H₂₉NO₅
MW: 423.51 g/mol
MDL Number: MFCD01862868
CAS Number: 957509-29-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR