Fmoc-Nd-trityl-L-β-homoglutamine
Supplier: Matrix Scientific
Synonyms:
Fmoc-β-HomoGln(Trt)-OH, Fmoc-β-HoGln(Trt)-OH
042375-250MG
042375-1G
101855-556EA
300
USD
101855-556
101855-558
Fmoc-Nd-trityl-L-β-homoglutamine
Fmoc-Nd-trityl-L-beta-homoglutamine
Hazard: Irritant
Formula:
C₄₀H₃₆N₂O₅ |
MDL Number:
MFCD03427580 CAS Number: 401915-55-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...