Fmoc-L-Nw-boc-β-homolysine

Supplier: Matrix Scientific

Synonyms: Fmoc-β-HomoLys(Boc)-OH

042374-250MG 042374-1G
101855-552EA 275.38 USD
101855-552 101855-554
Fmoc-L-Nw-boc-β-homolysine
Fmoc-L-Nw-boc-beta-homolysine

Hazard: Irritant

Formula: C₂₇H₃₄N₂O₆
MW: 482.58 g/mol
MDL Number: MFCD03002713
CAS Number: 203854-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR