Fmoc-L-β-homovaline

Supplier: Matrix Scientific

Synonyms: Fmoc-β-HoVal-OH, Fmoc-β-HomoVal-OH

042373-1G 042373-5G
101855-548EA 661.54 USD
101855-548 101855-550
Fmoc-L-β-homovaline
Fmoc-L-beta-homovaline

Hazard: Irritant

Formula: C₂₁H₂₃NO₄
MW: 353.42 g/mol
MDL Number: MFCD01862853
CAS Number: 172695-33-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR