Fmoc-L-β-homoproline

Supplier: Matrix Scientific

Synonyms: Fmoc-β-HomoPro-OH, Fmoc-β-HoPro-OH

042371-250MG 042371-1G
101855-540EA 238.46 USD
101855-540 101855-542
Fmoc-L-β-homoproline
Fmoc-L-beta-homoproline

Hazard: Irritant

Formula: C₂₁H₂₁NO₄
MW: 351.41 g/mol
MDL Number: MFCD01863058
CAS Number: 193693-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR