Fmoc-L-β-homophenylalanine

Supplier: Matrix Scientific

Synonyms: Fmoc-B-HoPhe-OH, Fmoc-β-HoPhe-OH, Fmoc-β-HomoPhe-OH

042370-250MG 042370-1G
101855-536EA 238.46 USD
101855-536 101855-538
Fmoc-L-β-homophenylalanine
Fmoc-L-beta-homophenylalanine

Hazard: Irritant

Formula: C₂₅H₂₃NO₄
MDL Number: MFCD01863055
CAS Number: 193954-28-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR