Fmoc-L-β-homoalanine

Supplier: Matrix Scientific

Synonyms: Fmoc-β-HoAla-OH, Fmoc-β-HomoAla-OH

042364-1G 042364-5G
101855-512EA 686.15 USD
101855-512 101855-514
Fmoc-L-β-homoalanine
Fmoc-L-beta-homoalanine

Hazard: Irritant

Formula: C₁₉H₁₉NO₄
MW: 325.37 g/mol
MDL Number: MFCD00270346
CAS Number: 193954-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR