Fmoc-γ-trityl-L-β-homoasparagine

Supplier: Matrix Scientific

042363-250MG 042363-1G
101855-508EA 386.15 USD
101855-508 101855-510
Fmoc-γ-trityl-L-β-homoasparagine
Fmoc-gamma-trityl-L-beta-homoasparagine

Hazard: Irritant

Formula: C₃₉H₃₄N₂O₅
MDL Number: MFCD03427579
CAS Number: 283160-20-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR