Fmoc-DL-β-leucine

Supplier: Matrix Scientific

Synonyms: 3-(Fmoc-amino)-4-methylpentanoic acid

042362-1G 042362-5G
101855-504EA 1233.85 USD
101855-504 101855-506
Fmoc-DL-β-leucine
Fmoc-DL-beta-leucine

Hazard: Irritant

Formula: C₂₁H₂₃NO₄
MW: 353.42 g/mol
MDL Number: MFCD01863209
CAS Number: 282524-93-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR