N-Fmoc-β-alanine

Supplier: Matrix Scientific

Synonyms: Fmoc-β-Ala-OH P, Fmoc-β-Ala-p-alkoxybenzyl alcohol, N-Fmoc-beta-alanine, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-beta-alanine

042360-25G 042360-100G 042360-5G
101855-496EA 129.23 USD
101855-496 101855-498 102551-306
N-Fmoc-β-alanine
N-Fmoc-β-alanine

Hazard: Irritant

Formula: C₁₈H₁₇NO₄
MW: 311.3 g/mol
Melting Pt: 142-147°C
MDL Number: MFCD00063328
CAS Number: 35737-10-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR