Fmoc-3-amino-3-(3-bromophenyl)propionic acid

Supplier: Matrix Scientific

042356-1G 042356-5G
101855-486EA 661.54 USD
101855-486 101855-488
Fmoc-3-amino-3-(3-bromophenyl)propionic acid
Fmoc-3-amino-3-(3-bromophenyl)propionic acid

Hazard: Irritant

Formula: C₂₄H₂₀BrNO₄
MDL Number: MFCD02090653
CAS Number: 276262-71-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR