Fmoc-(S)-3-amino-4-(4-bromophenyl)butyric acid

Supplier: Matrix Scientific

Synonyms: (S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(4-bromophenyl)butanoic acid

042354-1G 042354-5G
101855-478EA 823.08 USD
101855-478 101855-480
Fmoc-(S)-3-amino-4-(4-bromophenyl)butyric acid
Fmoc-(S)-3-amino-4-(4-bromophenyl)butyric acid

Hazard: Irritant

Formula: C₂₅H₂₂BrNO₄
MW: 480.37 g/mol
MDL Number: MFCD01861016
CAS Number: 270062-86-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR